siRNA Details
| Gene Symbol: Kcns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031778.2 | NP_113966.2 | 3 | 1324 | ||
| GenBank mRNA | AF029056.1 | - | 1 | 1022 | ||
| Y17607.1 | - | 3 | 1324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcns3| Gene Symbol: Kcns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031778.2 | NP_113966.2 | 3 | 1324 | ||
| GenBank mRNA | AF029056.1 | - | 1 | 1022 | ||
| Y17607.1 | - | 3 | 1324 | |||