siRNA Details
| Gene Symbol: Apba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031779.2 | NP_113967.1 | 5 | 1820 | ||
| XM_008760312.1 | XP_008758534.1 | 4 | 1953 | |||
| GenBank mRNA | AF029105.1 | - | 2040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apba1| Gene Symbol: Apba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031779.2 | NP_113967.1 | 5 | 1820 | ||
| XM_008760312.1 | XP_008758534.1 | 4 | 1953 | |||
| GenBank mRNA | AF029105.1 | - | 2040 | |||