siRNA Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031781.1 | NP_113969.1 | 5 | 1046 | ||
| XM_006241018.2 | XP_006241080.1 | 6 | 1207 | |||
| XM_006241019.2 | XP_006241081.1 | 6 | 1180 | |||
| XM_006241020.2 | XP_006241082.1 | 6 | 1205 | |||
| GenBank mRNA | AF029109.1 | - | 5 | 1046 | ||
| BC161842.1 | - | 6 | 1108 | 9102177 | ||

Apba3
Genomic Map