siRNA Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031781.1 | NP_113969.1 | 9, 10 | 1717 | ||
| XM_006241020.2 | XP_006241082.1 | 10, 11 | 1876 | |||
| GenBank mRNA | AF029109.1 | - | 9, 10 | 1717 | ||
| BC161842.1 | - | 10, 11 | 1779 | 9102177 | ||

Apba3
Genomic Map