siRNA Details
| Gene Symbol: Slc32a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031782.1 | NP_113970.1 | 2 | 601 | ||
| XM_006235441.2 | XP_006235503.1 | 2 | 765 | |||
| GenBank mRNA | AF030253.1 | - | 2 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc32a1| Gene Symbol: Slc32a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031782.1 | NP_113970.1 | 2 | 601 | ||
| XM_006235441.2 | XP_006235503.1 | 2 | 765 | |||
| GenBank mRNA | AF030253.1 | - | 2 | 601 | ||