siRNA Details
| Gene Symbol: STYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130701.1 | NP_001124173.1 | 11 | 649 | ||
| NM_145251.3 | NP_660294.1 | 10 | 897 | |||
| XM_006720242.3 | XP_006720305.1 | 8 | 517 | |||
| XM_011537108.2 | XP_011535410.1 | 10 | 564 | |||
| XM_011537109.2 | XP_011535411.1 | 7 | 458 | |||
| GenBank mRNA | AK098195.1 | - | 10 | 880 | ||
| AL832591.1 | - | 11 | 634 | |||
| AL832697.1 | - | 1 | 37 | |||
| BC020265.2 | - | 10 | 848 | 3922829 | ||
| BC146995.1 | - | 10 | 787 | 9056923 | ||
| BC146998.1 | - | 10 | 787 | 9056926 | ||
| BX647636.1 | - | 10 | 855 | |||

STYX
Genomic Map