siRNA Details
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031786.1 | NP_113974.1 | 2 | 296 | ||
GenBank mRNA | AF036255.1 | - | 2 | 296 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031786.1 | NP_113974.1 | 2 | 296 | ||
GenBank mRNA | AF036255.1 | - | 2 | 296 |