siRNA Details
| Gene Symbol: Hipk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031787.1 | NP_113975.1 | 2 | 1165 | ||
| XM_006234658.2 | XP_006234720.1 | 2 | 1306 | |||
| XM_008762072.1 | XP_008760294.1 | 2 | 1308 | |||
| GenBank mRNA | AF036959.1 | - | 2 | 1165 | ||
| BC100068.1 | - | 2 | 815 | 7111326 | ||

Hipk3
Genomic Map