siRNA Details
| Gene Symbol: STYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130701.1 | NP_001124173.1 | 4 | 185 | ||
| NM_145251.3 | NP_660294.1 | 3 | 433 | |||
| XM_011537108.2 | XP_011535410.1 | 3 | 100 | |||
| GenBank mRNA | AK098195.1 | - | 3 | 416 | ||
| AL832591.1 | - | 4 | 170 | |||
| BC020265.2 | - | 3 | 384 | 3922829 | ||
| BC146995.1 | - | 3 | 323 | 9056923 | ||
| BC146998.1 | - | 3 | 323 | 9056926 | ||
| BX647636.1 | - | 3 | 391 | |||

STYX
Genomic Map