siRNA Details
| Gene Symbol: Cd82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031797.2 | NP_113985.2 | 7 | 622 | ||
| XM_006234597.2 | XP_006234659.1 | 7 | 696 | |||
| XM_006234598.1 | XP_006234660.1 | 7 | 510 | |||
| XM_008762039.1 | XP_008760261.1 | 6 | 415 | |||
| GenBank mRNA | AF049882.1 | - | 7 | 536 | ||
| BC072498.1 | - | 7 | 622 | 7096826 | ||

Cd82
Genomic Map