siRNA Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031800.1 | NP_113988.1 | 1 | 185 | ||
| XM_006250269.2 | XP_006250331.1 | 3 | 250 | |||
| XM_006250270.2 | XP_006250332.1 | 1 | 200 | |||
| XM_006250272.2 | XP_006250334.1 | 3 | 330 | |||
| XM_006250273.2 | XP_006250335.1 | 2 | 256 | |||
| XM_006250274.2 | XP_006250336.1 | 1 | 204 | |||
| GenBank mRNA | AF053362.1 | - | 1 | 185 | ||

Dedd
Genomic Map