siRNA Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031801.1 | NP_113989.1 | 12 | 1709 | ||
| XM_006230582.2 | XP_006230644.1 | 12 | 2577 | |||
| XM_006230583.2 | XP_006230645.1 | 11 | 2487 | |||
| XM_008760011.1 | XP_008758233.1 | 12 | 1628 | |||
| XM_008760012.1 | XP_008758234.1 | 12 | 1625 | |||
| XM_008760013.1 | XP_008758235.1 | 11 | 1538 | |||
| XM_008760014.1 | XP_008758236.1 | 12 | 2593 | |||
| XM_008760015.1 | XP_008758237.1 | 7 | 1221 | |||
| XM_008760016.1 | XP_008758238.1 | 7 | 1218 | |||
| GenBank mRNA | AF055884.1 | - | 12 | 1709 | ||
| U59659.1 | - | 12 | 1455 | |||

Deaf1
Genomic Map