siRNA Details
| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031808.1 | NP_113996.1 | 8 | 1275 | ||
| GenBank mRNA | AF067793.1 | - | 8 | 1275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn6| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031808.1 | NP_113996.1 | 8 | 1275 | ||
| GenBank mRNA | AF067793.1 | - | 8 | 1275 | ||