siRNA Details
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031811.3 | NP_113999.2 | 4 | 394 | ||
GenBank mRNA | AF069306.1 | - | 4 | 380 | ||
BC059126.1 | - | 4 | 398 | 6889440 | ||
FQ213939.1 | - | 4 | 406 | |||
FQ219168.1 | - | 4 | 420 |
