siRNA Details
| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031817.1 | NP_114005.1 | 2 | 1148 | ||
| XM_008771551.1 | XP_008769773.1 | 2 | 2043 | |||
| GenBank mRNA | AF104362.1 | - | 2 | 1148 | ||
| Gene Symbol: LOC103690116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771520.1 | XP_008769742.1 | 3965 | |||
| XM_008771521.1 | XP_008769743.1 | 2043 | ||||

Omd
Genomic Map