siRNA Details
| Gene Symbol: Clic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031818.1 | NP_114006.1 | 3 | 231 | ||
| GenBank mRNA | AF104119.1 | - | 3 | 231 | ||
| EF397567.1 | - | 3 | 231 | |||
| FQ222737.1 | - | 341 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clic4| Gene Symbol: Clic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031818.1 | NP_114006.1 | 3 | 231 | ||
| GenBank mRNA | AF104119.1 | - | 3 | 231 | ||
| EF397567.1 | - | 3 | 231 | |||
| FQ222737.1 | - | 341 | ||||