siRNA Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031820.1 | NP_114008.1 | 7 | 740 | ||
| XM_006239598.2 | XP_006239660.1 | 7 | 1054 | |||
| XM_008764388.1 | XP_008762610.1 | 7 | 1055 | |||
| XM_008764389.1 | XP_008762611.1 | 7 | 1055 | |||
| XM_008764390.1 | XP_008762612.1 | 7 | 1055 | |||
| XM_008764391.1 | XP_008762613.1 | 8 | 756 | |||
| GenBank mRNA | AF143545.2 | - | 7 | 740 | ||
| AF143546.2 | - | 7 | 740 | |||
| BC062070.1 | - | 5 | 837 | |||

Dvl1
Genomic Map