siRNA Details
| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031822.1 | NP_114010.1 | 9 | 2274 | ||
| XM_006237744.2 | XP_006237806.1 | 11 | 2772 | |||
| XM_006237745.2 | XP_006237807.1 | 11 | 2375 | |||
| XM_006237747.2 | XP_006237809.1 | 10 | 3464 | |||
| XM_008763516.1 | XP_008761738.1 | 10 | 3657 | |||
| GenBank mRNA | AF136943.1 | - | 9 | 2274 | ||

Ncoa2
Genomic Map