siRNA Details
| Gene Symbol: Ctcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031824.1 | NP_114012.1 | 4 | 983 | ||
| XM_006255536.1 | XP_006255598.1 | 3 | 933 | |||
| XM_008772536.1 | XP_008770758.1 | 2 | 950 | |||
| XM_008772537.1 | XP_008770759.1 | 2 | 950 | |||
| XM_008772538.1 | XP_008770760.1 | 2 | 950 | |||
| XM_008772539.1 | XP_008770761.1 | 2 | 950 | |||
| GenBank mRNA | AF133731.2 | - | 4 | 1019 | ||

Ctcf
Genomic Map