siRNA Details
| Gene Symbol: Vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031827.1 | NP_114015.1 | 2 | 250 | ||
| GenBank mRNA | AF132812.1 | - | 2 | 250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vamp8| Gene Symbol: Vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031827.1 | NP_114015.1 | 2 | 250 | ||
| GenBank mRNA | AF132812.1 | - | 2 | 250 | ||