siRNA Details
| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270800.1 | NP_001257729.1 | 5, 6 | 569 | ||
| NM_001270801.1 | NP_001257730.1 | 5, 6 | 578 | |||
| NM_031830.2 | NP_114018.1 | 5, 6 | 569 | |||
| XM_006246952.1 | XP_006247014.1 | 6, 7 | 1190 | |||
| XM_008768119.1 | XP_008766341.1 | 4, 5 | 556 | |||
| GenBank mRNA | AF023302.1 | - | 5, 6 | 548 | ||
| AF023303.1 | - | 5, 6 | 548 | |||
| AF023304.1 | - | 4, 5 | 466 | |||
| BC083550.1 | - | 4, 5 | 445 | 7133999 | ||
| CB581049.1 | - | 5, 6 | 569 | |||
| U64999.1 | - | 2, 3 | 421 | |||

Flot2
Genomic Map