siRNA Details
| Gene Symbol: Rtn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031831.1 | NP_114019.1 | 3 | 2262 | ||
| XM_006251605.2 | XP_006251667.1 | 3 | 2448 | |||
| XM_006251606.1 | XP_006251668.1 | 3 | 2448 | |||
| XM_008770468.1 | XP_008768690.1 | 2 | 3849 | |||
| GenBank mRNA | AJ242961.1 | - | 3 | 2262 | ||

Rtn4
Genomic Map