siRNA Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031835.2 | NP_114023.1 | 12 | 1271 | ||
| XM_006232069.2 | XP_006232131.1 | 12 | 1339 | |||
| GenBank mRNA | AB002584.1 | - | 12 | 1242 | ||
| BC081765.1 | - | 12 | 1271 | 7106995 | ||
| D38100.1 | - | 12 | 1286 | |||
| FQ219625.1 | - | 4 | 352 | |||

Agxt2
Genomic Map