siRNA Details
| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031839.3 | NP_114027.2 | 8 | 1222 | ||
| GenBank mRNA | BC078707.1 | - | 8 | 1222 | 7110580 | |
| FQ209508.1 | - | 6 | 821 | |||
| FQ218733.1 | - | 3 | 349 | |||
| FQ218882.1 | - | 5 | 571 | |||
| U04733.1 | - | 8 | 1203 | |||

Cyp2c23
Genomic Map