siRNA Details
| Gene Symbol: Clta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031974.1 | NP_114180.1 | 4 | 517 | ||
| XM_006238104.1 | XP_006238166.1 | 4 | 1290 | |||
| XM_006238105.1 | XP_006238167.1 | 4 | 1290 | |||
| XM_006238106.1 | XP_006238168.1 | 4 | 1290 | |||
| GenBank mRNA | BC087577.1 | - | 4 | 550 | 7307624 | |
| FQ213034.1 | - | 4 | 525 | |||
| FQ213766.1 | - | 4 | 518 | |||
| FQ220908.1 | - | 4 | 525 | |||
| FQ230032.1 | - | 4 | 518 | |||
| M15882.1 | - | 4 | 517 | |||
| M19260.1 | - | 4 | 517 | |||
| M19261.1 | - | 4 | 517 | |||

Clta
Genomic Map