siRNA Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031977.1 | NP_114183.1 | 4, 5 | 467 | ||
| XM_008762387.1 | XP_008760609.1 | 4, 5 | 476 | |||
| XM_008762388.1 | XP_008760610.1 | 4, 5 | 513 | |||
| XM_008762389.1 | XP_008760611.1 | 4, 5 | 584 | |||
| GenBank mRNA | AF130457.1 | - | 3, 4 | 449 | ||
| AF157016.1 | - | 4, 5 | 467 | |||
| DQ120509.1 | - | 3, 4 | 433 | |||
| DQ120510.1 | - | 3, 4 | 433 | |||

Src
Genomic Map