siRNA Details
| Gene Symbol: Ugt2b15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004271.1 | NP_001004271.1 | 1 | 404 | ||
| XM_008770060.1 | XP_008768282.1 | 1 | 431 | |||
| GenBank mRNA | BC078782.1 | - | 1 | 404 | 7107952 | |
| U06273.1 | - | 1 | 385 | |||
| U06274.1 | - | 1 | 386 | |||

Ugt2b15
Genomic Map