siRNA Details
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172009.1 | NP_742006.1 | 2 | 1524 | ||
GenBank mRNA | S50879.1 | - | 2 | 1524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172009.1 | NP_742006.1 | 2 | 1524 | ||
GenBank mRNA | S50879.1 | - | 2 | 1524 |