siRNA Details
| Gene Symbol: Sycp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130735.1 | NP_570091.1 | 36 | 3910 | ||
| XM_008762546.1 | XP_008760768.1 | 25 | 2910 | |||
| XM_008762547.1 | XP_008760769.1 | 36 | 4856 | |||
| GenBank mRNA | Y08981.1 | - | 36 | 3910 | ||

Sycp2
Genomic Map