siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 10 | 1058 | ||
| XM_008761004.1 | XP_008759226.1 | 10 | 2960 | |||
| XM_008761005.1 | XP_008759227.1 | 10 | 1986 | |||
| XM_008761006.1 | XP_008759228.1 | 10 | 2963 | |||
| XM_008761007.1 | XP_008759229.1 | 10 | 1070 | |||
| GenBank mRNA | CB586027.1 | - | 3 | 236 | ||
| FQ211799.1 | - | 10 | 1045 | |||
| U78857.1 | - | 10 | 1048 | |||

Dclk1
Genomic Map