siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 12 | 1274 | ||
| XM_008761004.1 | XP_008759226.1 | 12 | 3176 | |||
| XM_008761007.1 | XP_008759229.1 | 12 | 1286 | |||
| GenBank mRNA | U78857.1 | - | 12 | 1264 | ||

Dclk1
Genomic Map