siRNA Details
| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031984.2 | NP_114190.1 | 9 | 696 | ||
| GenBank mRNA | BC081764.1 | - | 9 | 679 | 7110962 | |
| X04280.1 | - | 9 | 711 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calb1| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031984.2 | NP_114190.1 | 9 | 696 | ||
| GenBank mRNA | BC081764.1 | - | 9 | 679 | 7110962 | |
| X04280.1 | - | 9 | 711 | |||