siRNA Details
| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031984.2 | NP_114190.1 | 10, 11 | 795 | ||
| GenBank mRNA | BC081764.1 | - | 10, 11 | 778 | 7110962 | |
| FQ214326.1 | - | 9, 10 | 758 | |||
| X04280.1 | - | 10, 11 | 810 | |||

Calb1
Genomic Map