siRNA Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032059.1 | NP_114448.1 | 18 | 1887 | ||
| GenBank mRNA | AB020615.1 | - | 10 | 1014 | ||
| EU517502.1 | - | 18 | 1719 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkci| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032059.1 | NP_114448.1 | 18 | 1887 | ||
| GenBank mRNA | AB020615.1 | - | 10 | 1014 | ||
| EU517502.1 | - | 18 | 1719 | |||