siRNA Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032066.1 | NP_114455.1 | 5 | 438 | ||
| XM_006234599.2 | XP_006234661.1 | 5 | 605 | |||
| GenBank mRNA | BC061543.1 | - | 5 | 459 | 6888949 | |
| FQ213557.1 | - | 5 | 474 | |||
| FQ214777.1 | - | 5 | 503 | |||
| U81186.1 | - | 5 | 438 | |||

Hsd17b12
Genomic Map