siRNA Details
| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761978.1 | XP_008760200.1 | 252 | 59554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttn| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761978.1 | XP_008760200.1 | 252 | 59554 | ||