siRNA Details
| Gene Symbol: Synj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113371.1 | NP_001106842.1 | 22 | 3242 | ||
| NM_001113372.1 | NP_001106843.1 | 22 | 3242 | |||
| NM_032071.2 | NP_114460.1 | 22 | 3297 | |||
| GenBank mRNA | AY034050.1 | - | 22 | 3242 | ||
| AY034051.1 | - | 22 | 3242 | |||
| CK653289.1 | - | 11 | ||||
| U90312.1 | - | 22 | 3297 | |||

Synj2
Genomic Map