siRNA Details
| Gene Symbol: Irs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032074.1 | NP_114463.1 | 2 | 1369 | ||
| GenBank mRNA | BC161840.1 | - | 2 | 1221 | 9027966 | |
| U93880.1 | - | 2 | 1369 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irs3| Gene Symbol: Irs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032074.1 | NP_114463.1 | 2 | 1369 | ||
| GenBank mRNA | BC161840.1 | - | 2 | 1221 | 9027966 | |
| U93880.1 | - | 2 | 1369 | |||