siRNA Details
| Gene Symbol: Ghsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032075.3 | NP_114464.1 | 1 | 52 | ||
| GenBank mRNA | AB001982.1 | - | 1 | 52 | ||
| U94321.1 | - | 1 | 52 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ghsr| Gene Symbol: Ghsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032075.3 | NP_114464.1 | 1 | 52 | ||
| GenBank mRNA | AB001982.1 | - | 1 | 52 | ||
| U94321.1 | - | 1 | 52 | |||
Set Membership: |
|