siRNA Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032076.3 | NP_114465.3 | 2 | 1303 | ||
| XM_006231996.2 | XP_006232058.1 | 3 | 3872 | |||
| GenBank mRNA | D28860.1 | - | 3 | 1483 | ||
| U94709.1 | - | 2 | 1303 | |||
More Information
Set Membership: |
|

Ptger4
Genomic Map