siRNA Details
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032081.1 | NP_114470.1 | 12 | 1339 | ||
GenBank mRNA | FQ232772.1 | - | 12 | 1339 | ||
X61925.1 | - | 12 | 1339 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032081.1 | NP_114470.1 | 12 | 1339 | ||
GenBank mRNA | FQ232772.1 | - | 12 | 1339 | ||
X61925.1 | - | 12 | 1339 |