siRNA Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032083.1 | NP_114472.1 | 7 | 1362 | ||
| XM_006234386.2 | XP_006234448.1 | 13 | 2078 | |||
| XM_006234388.2 | XP_006234450.1 | 13 | 1318 | |||
| XM_006234389.2 | XP_006234451.1 | 13 | 2021 | |||
| XM_008761932.1 | XP_008760154.1 | 10 | 2078 | |||
| GenBank mRNA | BC094519.1 | - | 7 | 1411 | 7315609 | |
| X67250.1 | - | 7 | 1362 | |||

Chn1
Genomic Map