siRNA Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032083.1 | NP_114472.1 | 1 | 471 | ||
| GenBank mRNA | BC094519.1 | - | 1 | 520 | 7315609 | |
| X67250.1 | - | 1 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chn1| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032083.1 | NP_114472.1 | 1 | 471 | ||
| GenBank mRNA | BC094519.1 | - | 1 | 520 | 7315609 | |
| X67250.1 | - | 1 | 471 | |||