siRNA Details
| Gene Symbol: Enpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057104.2 | NP_476445.2 | 5 | 770 | ||
| GenBank mRNA | BC081747.1 | - | 5 | 730 | 7115236 | |
| D28560.1 | - | 6 | 764 | |||
| DQ131564.1 | - | 5 | 770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enpp2| Gene Symbol: Enpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057104.2 | NP_476445.2 | 5 | 770 | ||
| GenBank mRNA | BC081747.1 | - | 5 | 730 | 7115236 | |
| D28560.1 | - | 6 | 764 | |||
| DQ131564.1 | - | 5 | 770 | |||