siRNA Details
| Gene Symbol: Dnmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053354.3 | NP_445806.3 | 5, 6 | 600 | ||
| GenBank mRNA | AB012214.1 | - | 5, 6 | 584 | ||
| D64060.1 | - | 5, 6 | 472 | |||
| DV214494.1 | - | 5, 6 | 566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnmt1| Gene Symbol: Dnmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053354.3 | NP_445806.3 | 5, 6 | 600 | ||
| GenBank mRNA | AB012214.1 | - | 5, 6 | 584 | ||
| D64060.1 | - | 5, 6 | 472 | |||
| DV214494.1 | - | 5, 6 | 566 | |||