siRNA Details
| Gene Symbol: Dnmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053354.3 | NP_445806.3 | 28 | 3070 | ||
| GenBank mRNA | AB012214.1 | - | 28 | 3057 | ||
| CA511167.1 | - | 1 | 84 | |||
| CA513509.1 | - | 2 | 294 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnmt1| Gene Symbol: Dnmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053354.3 | NP_445806.3 | 28 | 3070 | ||
| GenBank mRNA | AB012214.1 | - | 28 | 3057 | ||
| CA511167.1 | - | 1 | 84 | |||
| CA513509.1 | - | 2 | 294 | |||