siRNA Details
| Gene Symbol: Ikbkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053355.2 | NP_445807.2 | 19, 20 | 2093 | ||
| GenBank mRNA | AF115282.1 | - | 18, 19 | 2007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikbkb| Gene Symbol: Ikbkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053355.2 | NP_445807.2 | 19, 20 | 2093 | ||
| GenBank mRNA | AF115282.1 | - | 18, 19 | 2007 | ||