siRNA Details
| Gene Symbol: Atox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053359.2 | NP_445811.1 | 3 | 222 | ||
| GenBank mRNA | FQ220104.1 | - | 3 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atox1| Gene Symbol: Atox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053359.2 | NP_445811.1 | 3 | 222 | ||
| GenBank mRNA | FQ220104.1 | - | 3 | 223 | ||