siRNA Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053360.2 | NP_445812.1 | 6 | 965 | ||
| XM_006256924.2 | XP_006256986.2 | 6 | 907 | |||
| XM_006256929.2 | XP_006256991.2 | 4 | 733 | |||
| XM_006256931.1 | XP_006256993.1 | 8 | 1258 | |||
| GenBank mRNA | AF131867.1 | - | 4 | 521 | ||
| AF230519.1 | - | 6 | 777 | |||
| AF230520.1 | - | 6 | 631 | |||
| AF255888.1 | - | 6 | 883 | |||
| BC070877.1 | - | 6 | 965 | 7096160 | ||

Sh3kbp1
Genomic Map