siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 14, 15 | 1509 | ||
| GenBank mRNA | AF149284.1 | - | 14, 15 | 1509 | ||
| FQ221525.1 | - | 1272 | ||||
| FQ226283.1 | - | 5, 6 | 648 | |||
| FQ227003.1 | - | 2, 3 | 266 | |||
| FQ232404.1 | - | 2, 3 | 265 | |||
| FQ233045.1 | - | 10, 11 | 1141 | |||
| U09228.1 | - | 8, 9 | 928 | |||

Tcf4
Genomic Map